Gene Transcripts

Gene Symbol
MRPL21
Entrez Gene ID
219927
Gene Name
mitochondrial ribosomal protein L21
Gene Aliases
L21mt, MRP-L21
Chromosome Location
Chr. 11: 68891276 - 68903835 on Build GRCh38
UniGene ID
Hs.503047
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_181514.1 NP_852615.1 2 161
NM_181515.1 NP_852616.1 2 161
XM_005273823.4 XP_005273880.1 2 158
GenBank mRNA AK096756.1 - 2 161
AK301989.1 - 2 158
AV711404.1 - 2 87
BC055088.1 - 2 160 6457142
BM916618.1 - 2 113

View Full Product Details