Gene Transcripts

Gene Symbol
CCNL2
Entrez Gene ID
81669
Gene Name
cyclin L2
Gene Aliases
ANIA-6B, CCNM, CCNS, HCLA-ISO, HLA-ISO, PCEE, SB138
Chromosome Location
Chr. 1: 1385711 - 1399338 on Build GRCh38
UniGene ID
Hs.515704
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001320153.1 NP_001307082.1 8 1610
NM_001320155.1 NP_001307084.1 8 900
NM_030937.4 NP_112199.2 7 886
NR_135154.1 7 2824
XM_011542216.2 XP_011540518.1 7 961
XM_011542219.2 XP_011540521.1 7 770
XM_011542221.2 XP_011540523.1 6 601
XM_017002420.1 XP_016857909.1 8 2391
XM_017002421.1 XP_016857910.1 9 1274
XM_017002422.1 XP_016857911.1 8 1008
GenBank mRNA AF251294.1 - 6 1339
AK000685.1 - 2 426
AK026796.1 - 2 1085
AK027770.1 - 2 1855
AK056120.1 - 8 1582
AK057133.1 - 6 601
AK074112.1 - 7 2795
AK225758.1 - 7 881
AK292268.1 - 2 409
AK293451.1 - 4 364
AY037150.1 - 7 860
AY116620.1 - 8 889
BC071622.1 - 2 273

View Full Product Details