Gene Transcripts

Gene Symbol
SH2D4A
Entrez Gene ID
63898
Gene Name
SH2 domain containing 4A
Gene Aliases
PPP1R38, SH2A
Chromosome Location
Chr. 8: 19313443 - 19396218 on Build GRCh38
UniGene ID
Hs.303208
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001174159.1 NP_001167630.1 10 2180
NM_001174160.1 NP_001167631.1 9 1945
NM_022071.3 NP_071354.2 10 2330
XM_011544610.1 XP_011542912.1 9 2376
XM_017013729.1 XP_016869218.1 9 2098
GenBank mRNA AK024620.1 - 8 1599
AK024799.1 - 10 2208
BC067117.1 - 7 1539

View Full Product Details