Gene Transcripts

Gene Symbol
CARD10
Entrez Gene ID
29775
Gene Name
caspase recruitment domain family member 10
Gene Aliases
BIMP1, CARMA3
Chromosome Location
Chr. 22: 37490362 - 37519203 on Build GRCh38
UniGene ID
Hs.57973
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014550.3 NP_055365.2 14 2296
GenBank mRNA AK074906.1 - 11 1526
AK296176.1 - 2 566
AY028896.1 - 14 2333
AY032928.1 - 14 2296
BC032058.1 - 12 1921
BC113659.1 - 14 2314 8322711
BC117403.1 - 14 2308 40125954

View Full Product Details