Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NR_028393.1 | 6 | 1355 | ||
NR_028394.1 | 7 | 1483 | |||
NR_028395.1 | 7 | 1483 | |||
NR_028396.1 | 6 | 1355 | |||
NR_028397.1 | 6 | 1220 | |||
NR_028398.1 | 4 | 974 | |||
NR_028399.1 | 8 | 1590 | |||
NR_028400.1 | 7 | 1462 |