Gene Transcripts
- Gene Symbol
- KPTN
- Entrez Gene ID
- 11133
- Gene Name
- kaptin, actin binding protein
- Gene Aliases
- 2E4, MRT41
- Chromosome Location
- Chr. 19: 47475141 - 47486792 on Build GRCh38
- UniGene ID
- Hs.25441
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AF105369.1 |
- |
3, 4 |
422 |
|
View Full Product Details