Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001040439.1 | NP_001035529.1 | 31 | 5572 | |
NM_001318852.1 | NP_001305781.1 | 32 | 5630 | ||
NM_015133.4 | NP_055948.2 | 32 | 5627 | ||
XM_005255190.2 | XP_005255247.1 | 31 | 5585 | ||
XM_006720869.2 | XP_006720932.1 | 32 | 7060 | ||
XM_011522429.2 | XP_011520731.1 | 33 | 5654 | ||
XM_011522430.2 | XP_011520732.1 | 32 | 5636 | ||
XM_011522431.2 | XP_011520733.1 | 33 | 5627 | ||
XM_011522432.2 | XP_011520734.1 | 32 | 5609 | ||
XM_011522433.2 | XP_011520735.1 | 28 | 6277 | ||
XM_017023079.1 | XP_016878568.1 | 31 | 5612 | ||
XM_017023080.1 | XP_016878569.1 | 32 | 5603 | ||
XM_017023081.1 | XP_016878570.1 | 31 | 5102 | ||
GenBank mRNA | AB028989.2 | - | 32 | 5590 | |
BC087861.1 | - | 1 | 1421 | ||
BC150266.1 | - | 32 | 5590 | 9007222 |