Gene Transcripts

Gene Symbol
RALY
Entrez Gene ID
22913
Gene Name
RALY heterogeneous nuclear ribonucleoprotein
Gene Aliases
HNRPCL2, P542
Chromosome Location
Chr. 20: 33993652 - 34084884 on Build GRCh38
UniGene ID
Hs.136947
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007367.3 NP_031393.2 4 890
NM_016732.2 NP_057951.1 4 890
XM_005260334.4 XP_005260391.1 3 805
XM_005260336.4 XP_005260393.1 3 805
XM_011528694.2 XP_011526996.1 4 448
XM_011528695.2 XP_011526997.1 5 1000
XM_017027731.1 XP_016883220.1 3 805
GenBank mRNA AF148457.1 - 4 616
AK222915.1 - 4 576
AK226085.1 - 4 789
AK291054.1 - 4 461
AK291079.1 - 4 467
AK299766.1 - 4 631
BC103753.1 - 4 637 6182358
BC105018.1 - 4 485 8144021
L38696.1 - 4 494

View Full Product Details