Gene Transcripts

Gene Symbol
RGS6
Entrez Gene ID
9628
Gene Name
regulator of G-protein signaling 6
Gene Aliases
GAP
Chromosome Location
Chr. 14: 71932100 - 72632478 on Build GRCh38
UniGene ID
Hs.509872
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001204416.2 NP_001191345.1 18 3049
NM_001204419.2 NP_001191348.1 17 2938
NM_001204420.2 NP_001191349.1 16 2884
NM_001204423.1 NP_001191352.1 16 2760
NM_001204424.1 NP_001191353.1 18 3287
NM_004296.6 NP_004287.3 17 2995
NR_135235.1 19 3112
XM_011537393.2 XP_011535695.1 20 3456
XM_011537397.1 XP_011535699.1 18 2626
XM_011537407.2 XP_011535709.1 21 3542
XM_017021818.1 XP_016877307.1 20 2971
XM_017021819.1 XP_016877308.1 20 2887
XM_017021820.1 XP_016877309.1 18 2695
XM_017021821.1 XP_016877310.1 18 2990
XM_017021823.1 XP_016877312.1 18 2942
XM_017021824.1 XP_016877313.1 18 2905
XM_017021829.1 XP_016877318.1 18 2945
GenBank mRNA AF156932.1 - 16 2710
L40394.1 - 3 1634

View Full Product Details