Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000355.3 | NP_000346.2 | 9 | 1737 | |
NM_001184726.1 | NP_001171655.1 | 9 | 1656 | ||
GenBank mRNA | BC001176.1 | - | 9 | 1615 | |
BC011239.1 | - | 9 | 1535 | 4153362 | |
L02648.1 | - | 9 | 1541 | ||
M60396.1 | - | 9 | 1525 |