Gene Transcripts
- Gene Symbol
- IFT43
- Entrez Gene ID
- 112752
- Gene Name
- intraflagellar transport 43
- Gene Aliases
- C14orf179, CED3
- Chromosome Location
- Chr. 14: 75985753 - 76084073 on Build GRCh38
- UniGene ID
- Hs.532626
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_052873.2 |
NP_443105.2
|
3, 4 |
249 |
|
GenBank mRNA |
BC010436.1 |
- |
3, 4 |
220 |
|
View Full Product Details