Gene Transcripts

Gene Symbol
VPS13C
Entrez Gene ID
54832
Gene Name
vacuolar protein sorting 13 homolog C
Gene Aliases
PARK23
Chromosome Location
Chr. 15: 61852389 - 62060465 on Build GRCh38
UniGene ID
Hs.511668
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001018088.2 NP_001018098.1 31 3261
NM_017684.4 NP_060154.3 29 3132
NM_018080.3 NP_060550.2 29 3132
NM_020821.2 NP_065872.1 31 3261
XM_011521713.2 XP_011520015.1 31 3222
XM_011521714.2 XP_011520016.1 31 3222
GenBank mRNA AB037842.1 - 31 3244
AJ608770.1 - 29 3115
AJ608771.1 - 31 3244
AJ626860.1 - 29 3115
AJ626861.1 - 31 3244
AK025364.1 - 26 2783
AK307518.1 - 29 3057

View Full Product Details