Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_015268.3 | NP_056083.3 | 22 | 2385 | |
XM_005247245.4 | XP_005247302.1 | 23 | 2582 | ||
XM_017006036.1 | XP_016861525.1 | 9 | 842 | ||
GenBank mRNA | AB014578.2 | - | 22 | 2434 | |
AK127112.1 | - | 6 | 473 | ||
AY369172.1 | - | 4 | 307 | ||
AY779857.1 | - | 22 | 2385 | ||
BC151246.1 | - | 23 | 2455 | 9007477 |