Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001100409.2 | NP_001093879.1 | 4 | 1336 | |
NM_001304792.1 | NP_001291721.1 | 4 | 1336 | ||
NM_015571.3 | NP_056386.2 | 4 | 1336 | ||
XM_006715457.3 | XP_006715520.1 | 5 | 1090 | ||
XM_011535746.2 | XP_011534048.1 | 3 | 888 | ||
XM_011535747.2 | XP_011534049.1 | 3 | 1242 | ||
XM_017010719.1 | XP_016866208.1 | 4 | 981 | ||
XM_017010720.1 | XP_016866209.1 | 4 | 294 | ||
XM_017010721.1 | XP_016866210.1 | 4 | 302 | ||
XM_017010722.1 | XP_016866211.1 | 5 | 1109 | ||
XM_017010723.1 | XP_016866212.1 | 5 | 1089 | ||
XM_017010724.1 | XP_016866213.1 | 5 | 1464 | ||
XM_017010725.1 | XP_016866214.1 | 3 | 1242 | ||
GenBank mRNA | AB018340.2 | - | 4 | 364 | |
AF196304.1 | - | 4 | 320 | ||
AF306508.1 | - | 4 | 335 | ||
AF307849.1 | - | 4 | 395 | ||
AK096455.1 | - | 4 | 918 | ||
AK126921.1 | - | 4 | 1336 | ||
BC028583.1 | - | 4 | 543 | 4838569 |