Gene Transcripts

Gene Symbol
SLC16A13
Entrez Gene ID
201232
Gene Name
solute carrier family 16 member 13
Gene Aliases
MCT13
Chromosome Location
Chr. 17: 7036075 - 7040121 on Build GRCh38
UniGene ID
Hs.448010
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_201566.2 NP_963860.1 4 1534
GenBank mRNA BC109286.1 - 4 1222
BC131739.1 - 4 1222
BC142618.1 - 4 1534 40147405

View Full Product Details