Gene Transcripts
- Gene Symbol
- Raly
- Entrez Gene ID
- 19383
- Gene Name
- hnRNP-associated with lethal yellow
- Gene Aliases
- AI663842, Merc
- Chromosome Location
- Chr. 2: 154791097 - 154871010 on Build GRCm38
- UniGene ID
- Mm.221440
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001139511.1 |
NP_001132983.1
|
7, 8 |
1351 |
|
NM_001139512.1 |
NP_001132984.1
|
8, 9 |
1117 |
|
NM_001139513.1 |
NP_001132985.1
|
8, 9 |
1399 |
|
NM_023130.3 |
NP_075619.2
|
8, 9 |
1434 |
|
GenBank mRNA |
AK014356.1 |
- |
8, 9 |
1187 |
|
AK135386.1 |
- |
7, 8 |
1110 |
|
AK152290.1 |
- |
8, 9 |
1191 |
|
AK154787.1 |
- |
7, 8 |
1108 |
|
AK168781.1 |
- |
8, 9 |
1191 |
|
BC004851.1 |
- |
7, 8 |
1095 |
2810841 |
BC016587.1 |
- |
8, 9 |
1183 |
2811315 |
L17076.1 |
- |
8, 9 |
1164 |
|
S72641.1 |
- |
8, 9 |
1385 |
|
View Full Product Details