Gene Transcripts

Gene Symbol
Plcxd3
Entrez Gene ID
239318
Gene Name
phosphatidylinositol-specific phospholipase C, X domain containing 3
Gene Aliases
B130016O10Rik
Chromosome Location
Chr. 15: 4375491 - 4575585 on Build GRCm38
UniGene ID
Mm.308996
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK038575.1 - 3 1595
BC137637.1 - 2 1359 8860656
BC137639.1 - 2 1359 8860658

View Full Product Details