Gene Transcripts

Gene Symbol
Sdc2
Entrez Gene ID
15529
Gene Name
syndecan 2
Gene Aliases
4833414L08Rik, AA960457, Hspg1, Synd2, syndecan-2
Chromosome Location
Chr. 15: 32920723 - 33034721 on Build GRCm38
UniGene ID
Mm.234266
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008304.2 NP_032330.1 5 1676
GenBank mRNA AK011042.1 - 5 1525
BC047144.1 - 5 1542 6395969
U00674.1 - 5 1676

View Full Product Details