Gene Transcripts
- Gene Symbol
- Aup1
- Entrez Gene ID
- 11993
- Gene Name
- ancient ubiquitous protein 1
- Gene Aliases
- AA589454
- Chromosome Location
- Chr. 6: 83054521 - 83057682 on Build GRCm38
- UniGene ID
- Mm.386752
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001301649.1 |
NP_001288578.1
|
5, 6 |
775 |
|
NM_007517.4 |
NP_031543.2
|
6, 7 |
926 |
|
GenBank mRNA |
AK154707.1 |
- |
6, 7 |
795 |
|
AK158035.1 |
- |
6, 7 |
717 |
|
AK167335.1 |
- |
6, 7 |
716 |
|
AK171011.1 |
- |
6, 7 |
729 |
|
AK171184.1 |
- |
6, 7 |
722 |
|
AK171202.1 |
- |
6, 7 |
736 |
|
AK171249.1 |
- |
6, 7 |
716 |
|
AK171541.1 |
- |
6, 7 |
716 |
|
AK171621.1 |
- |
6, 7 |
709 |
|
AK187544.1 |
- |
2 |
116 |
|
BC012699.1 |
- |
5, 6 |
559 |
3986517 |
BC016485.1 |
- |
6, 7 |
719 |
3962982 |
U41736.1 |
- |
6, 7 |
724 |
|
View Full Product Details