Gene Transcripts

Gene Symbol
Cdc25c
Entrez Gene ID
12532
Gene Name
cell division cycle 25C
Gene Aliases
Cdc25
Chromosome Location
Chr. 18: 34732993 - 34752130 on Build GRCm38
UniGene ID
Mm.286602
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK141737.1 - 3, 4 540
AK168287.1 - 3, 4 535
BC004669.1 - 2, 3 244 3498846
L16926.1 - 2, 3 668
U15562.1 - 3, 4 506

View Full Product Details