Gene Transcripts

Gene Symbol
Tspan17
Entrez Gene ID
74257
Gene Name
tetraspanin 17
Gene Aliases
2210021G21Rik, AI047581, Fbxo23, Tm4sf17
Chromosome Location
Chr. 13: 54789352 - 54796896 on Build GRCm38
UniGene ID
Mm.32883
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028841.3 NP_083117.2 8 1404
GenBank mRNA AK008761.1 - 8 1349
AK171691.1 - 8 1353
BC010346.1 - 8 1349 2811935

View Full Product Details