Gene Transcripts

Gene Symbol
Mrpl22
Entrez Gene ID
216767
Gene Name
mitochondrial ribosomal protein L22
Gene Aliases
E030011D16Rik, HSPC158, MRP-L25, Rpml25
Chromosome Location
Chr. 11: 58171604 - 58179580 on Build GRCm38
UniGene ID
Mm.259907
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175001.3 NP_778166.2 7 506
GenBank mRNA AK077615.1 - 7 500
AK086913.1 - 7 485
AK164417.1 - 7 485
BC061191.1 - 7 496 6703228

View Full Product Details