Gene Transcripts

Gene Symbol
Lzts1
Entrez Gene ID
211134
Gene Name
leucine zipper, putative tumor suppressor 1
Gene Aliases
F37, FEZ1
Chromosome Location
Chr. 8: 69132669 - 69184220 on Build GRCm38
UniGene ID
Mm.409446
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_199364.2 NP_955396.2 2 668
GenBank mRNA AF288601.1 - 2 668
BC127606.1 - 2 667
BC127607.1 - 2 668 40057969
BC139350.1 - 2 724 8862372
BC139352.1 - 2 724 8862374

View Full Product Details