Gene Transcripts

Gene Symbol
Olfr796
Entrez Gene ID
258933
Gene Name
olfactory receptor 796
Gene Aliases
MOR269-1
Chromosome Location
Chr. 10: 129607547 - 129608479 on Build GRCm38
UniGene ID
Mm.377755
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146931.1 NP_667142.1 1 778

View Full Product Details