Gene Transcripts

Gene Symbol
Zc3h8
Entrez Gene ID
57432
Gene Name
zinc finger CCCH type containing 8
Gene Aliases
AU020882, E130108N08Rik, Fliz1, Zc3hdc8
Chromosome Location
Chr. 2: 128926268 - 128944084 on Build GRCm38
UniGene ID
Mm.11675
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020594.2 NP_065619.2 6 773
GenBank mRNA AF061961.1 - 6 781
AK132679.1 - 6 720
AK164491.1 - 6 720
BC048687.1 - 6 705 6334861

View Full Product Details