Gene Transcripts

Gene Symbol
Nkiras2
Entrez Gene ID
71966
Gene Name
NFKB inhibitor interacting Ras-like protein 2
Gene Aliases
2410003M04Rik, 4930527H08Rik, D630018G21Rik, KBRAS2
Chromosome Location
Chr. 11: 100622955 - 100627602 on Build GRCm38
UniGene ID
Mm.274734
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028024.2 NP_082300.1 4 855
GenBank mRNA AK010371.1 - 4 855
AK052645.1 - 4 855
AK083475.1 - 4 898
AK149696.1 - 4 999
AK169878.1 - 4 880
AK208908.1 - 1 173
BC013469.1 - 3 808 4219892
BC024398.1 - 3 798 5003001

View Full Product Details