Gene Transcripts

Gene Symbol
Fbxo45
Entrez Gene ID
268882
Gene Name
F-box protein 45
Gene Aliases
2610017J04Rik, AI463119, BC026799
Chromosome Location
Chr. 16: 32230112 - 32247025 on Build GRCm38
UniGene ID
Mm.256137
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173439.2 NP_775615.2 2 656
GenBank mRNA BC026799.1 - 2 728
BC088738.1 - 2 656 6337188
BC098203.1 - 2 733 5705872

View Full Product Details