Gene Transcripts

Gene Symbol
Ube2d1
Entrez Gene ID
216080
Gene Name
ubiquitin-conjugating enzyme E2D 1
Gene Aliases
UBCH5
Chromosome Location
Chr. 10: 71254980 - 71285262 on Build GRCm38
UniGene ID
Mm.270530
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145420.2 NP_663395.1 6 494
GenBank mRNA AK144468.1 - 6 449
AK148364.1 - 6 482
BC019464.1 - 6 465 4205941

View Full Product Details