Gene Transcripts

Gene Symbol
Parm1
Entrez Gene ID
231440
Gene Name
prostate androgen-regulated mucin-like protein 1
Gene Aliases
2210012L08Rik, 9130213B05Rik, W91666
Chromosome Location
Chr. 5: 91517700 - 91624011 on Build GRCm38
UniGene ID
Mm.5002
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145562.2 NP_663537.1 4 2028
GenBank mRNA AK161172.1 - 1991
AK167402.1 - 4 2017
BC006604.1 - 4 2029 3966312

View Full Product Details