Gene Transcripts

Gene Symbol
Fermt2
Entrez Gene ID
218952
Gene Name
fermitin family homolog 2 (Drosophila)
Gene Aliases
AA960555, Kindlin-2, Mig2, Plekhc1
Chromosome Location
Chr. 14: 45458792 - 45530113 on Build GRCm38
UniGene ID
Mm.210018
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146054.2 NP_666166.2 15 2807
GenBank mRNA AK145030.1 - 15 2750
BC033436.1 - 15 2728 4950810
BC034168.1 - 6 1461

View Full Product Details