Gene Transcripts

Gene Symbol
Rufy1
Entrez Gene ID
216724
Gene Name
RUN and FYVE domain containing 1
Gene Aliases
3000002E04Rik, Rabip4, ZFYVE12
Chromosome Location
Chr. 11: 50389286 - 50431111 on Build GRCm38
UniGene ID
Mm.390400
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172557.2 NP_766145.1 2 394
GenBank mRNA AJ250024.1 - 1 70
AK046625.1 - 2 411
AK046633.1 - 2 395
AK051178.1 - 2 400

View Full Product Details