Gene Transcripts

Gene Symbol
Ythdf1
Entrez Gene ID
228994
Gene Name
YTH domain family 1
Gene Aliases
2210410K23Rik, 8030473O16
Chromosome Location
Chr. 2: 180904377 - 180920936 on Build GRCm38
UniGene ID
Mm.30205
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173761.3 NP_776122.1 5 2995
GenBank mRNA AK152264.1 - 2 1361
AK168430.1 - 2 1605
BC065050.1 - 5 2995 6418891

View Full Product Details