Gene Transcripts

Gene Symbol
Enpp4
Entrez Gene ID
224794
Gene Name
ectonucleotide pyrophosphatase/phosphodiesterase 4
Gene Aliases
4933413N07Rik, AA986363, AI195364, E-NPP 4, Gm90, mKIAA0879
Chromosome Location
Chr. 17: 44096307 - 44105924 on Build GRCm38
UniGene ID
Mm.255971
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_199016.2 NP_950181.2 2 419
GenBank mRNA AK090098.1 - 2 420
AK157622.1 - 2 315
AK173048.1 - 2 420
BC027749.1 - 2 373 5065563

View Full Product Details