Gene Transcripts

Gene Symbol
Vmn1r32
Entrez Gene ID
171188
Gene Name
vomeronasal 1 receptor 32
Gene Aliases
V1rc15
Chromosome Location
Chr. 6: 66552183 - 66559708 on Build GRCm38
UniGene ID
Mm.222748
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_134170.3 NP_598931.1 3 2410
GenBank mRNA AK029205.1 - 3 2411

View Full Product Details