Gene Transcripts
- Gene Symbol
- Slc30a9
- Entrez Gene ID
- 109108
- Gene Name
- solute carrier family 30 (zinc transporter), member 9
- Gene Aliases
- 2310024J23Rik, AL024256, GAC63, HUEL
- Chromosome Location
- Chr. 5: 67306937 - 67356834 on Build GRCm38
- UniGene ID
- Mm.234455
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_178651.3 |
NP_848766.2
|
12, 13 |
1212 |
|
GenBank mRNA |
AF263460.1 |
- |
10, 11 |
766 |
|
AK028885.1 |
- |
12, 13 |
1179 |
|
AK033990.1 |
- |
|
237 |
|
AK035541.1 |
- |
12, 13 |
1199 |
|
AK038559.1 |
- |
11, 12 |
1118 |
|
AK082853.1 |
- |
12, 13 |
1214 |
|
AK143277.1 |
- |
12, 13 |
1213 |
|
AK149549.1 |
- |
4, 5 |
3663 |
|
AK179008.1 |
- |
4, 5 |
300 |
|
AK182342.1 |
- |
4, 5 |
290 |
|
AK204464.1 |
- |
3, 4 |
231 |
|
AK205884.1 |
- |
4, 5 |
258 |
|
AY682914.1 |
- |
13, 14 |
1205 |
|
BC023833.1 |
- |
4, 5 |
316 |
|
BC026565.1 |
- |
8, 9 |
630 |
|
BC027806.1 |
- |
12, 13 |
974 |
|
BC031705.1 |
- |
9, 10 |
726 |
|
BC055773.1 |
- |
9, 10 |
665 |
|
BC078440.1 |
- |
10, 11 |
789 |
|
BC141008.1 |
- |
13, 14 |
1178 |
9055620 |
View Full Product Details