Gene Transcripts

Gene Symbol
Sccpdh
Entrez Gene ID
109232
Gene Name
saccharopine dehydrogenase (putative)
Gene Aliases
AW214504, C330023F11Rik
Chromosome Location
Chr. 1: 179668231 - 179687184 on Build GRCm38
UniGene ID
Mm.136458
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178653.3 NP_848768.1 2 380
GenBank mRNA AK042919.1 - 2 268
AK046033.1 - 2 381
AK046468.1 - 2 367
AK050554.1 - 2 328
AK077695.1 - 2 330
AK081021.1 - 1 2389
AK082803.1 - 2 265
AK086448.1 - 2 403
AK086632.1 - 2 378
AK145399.1 - 2 364
AK166018.1 - 2 292
AK182882.1 - 2 122
AK215316.1 - 2 122
BC025803.1 - 2 377 5100552

View Full Product Details