Gene Transcripts

Gene Symbol
Memo1
Entrez Gene ID
76890
Gene Name
mediator of cell motility 1
Gene Aliases
0610016J10Rik, D930048L02Rik
Chromosome Location
Chr. 17: 74198398 - 74295544 on Build GRCm38
UniGene ID
Mm.311874
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133771.2 NP_598532.1 6 597
GenBank mRNA AF363447.1 - 7 659
BC013819.1 - 6 614 3489363

View Full Product Details