Gene Transcripts

Gene Symbol
Phkb
Entrez Gene ID
102093
Gene Name
phosphorylase kinase beta
Gene Aliases
AI463271
Chromosome Location
Chr. 8: 85840984 - 86064254 on Build GRCm38
UniGene ID
Mm.237296
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_199446.1 NP_955517.1 7, 8 730
GenBank mRNA AK089493.1 - 8, 9 768
AK165815.1 - 7, 8 725
BC053105.1 - 7, 8 730 6409425

View Full Product Details