Gene Transcripts
- Gene Symbol
- Ecd
- Entrez Gene ID
- 70601
- Gene Name
- ecdysoneless homolog (Drosophila)
- Gene Aliases
- 5730461K03Rik, Hsgt1
- Chromosome Location
- Chr. 14: 20319859 - 20348121 on Build GRCm38
- UniGene ID
- Mm.35105
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_027475.3 |
NP_081751.1
|
14 |
2780 |
|
GenBank mRNA |
AK028664.1 |
- |
14 |
2736 |
|
View Full Product Details