Gene Transcripts

Gene Symbol
Rbm19
Entrez Gene ID
74111
Gene Name
RNA binding motif protein 19
Gene Aliases
1200009A02Rik, AV302714, NPO
Chromosome Location
Chr. 5: 120116513 - 120198971 on Build GRCm38
UniGene ID
Mm.41022
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028762.1 NP_083038.1 14 1794
GenBank mRNA AK004657.1 - 5 609
AK146493.1 - 14 1827
AK167100.1 - 14 1791
BC025619.1 - 14 1794 5320947
BC034010.1 - 14 1760 4225239

View Full Product Details