Gene Transcripts

Gene Symbol
Ap5m1
Entrez Gene ID
74385
Gene Name
adaptor-related protein complex 5, mu 1 subunit
Gene Aliases
4932432K03Rik, Mu5, Mudeng
Chromosome Location
Chr. 14: 49066495 - 49094114 on Build GRCm38
UniGene ID
Mm.379235
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_144535.4 NP_653118.3 3 1049
GenBank mRNA AK016540.1 - 3 996
AK030928.1 - 3 976
AK031527.1 - 3 1448
AK041400.1 - 3 550
AK051372.1 - 3 996
AK154166.1 - 3 1002
AK164148.1 - 3 820
AK170232.1 - 3 826
BC023094.1 - 3 1049 4017596

View Full Product Details