Gene Transcripts

Gene Symbol
Fndc3b
Entrez Gene ID
72007
Gene Name
fibronectin type III domain containing 3B
Gene Aliases
1600019O04Rik, AW550168, Fad104, mKIAA4164
Chromosome Location
Chr. 3: 27416161 - 27711662 on Build GRCm38
UniGene ID
Mm.38832
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173182.2 NP_775274.2 6 697
GenBank mRNA AB098596.1 - 6 635
AK045099.1 - 6 792
AK147346.1 - 7 708
AK220297.1 - 6 697
BC067389.1 - 6 703 30431862
DQ192037.1 - 5 603

View Full Product Details