Gene Transcripts

Gene Symbol
Mcoln2
Entrez Gene ID
68279
Gene Name
mucolipin 2
Gene Aliases
3300002C04Rik, AI549968, C86638, TRPML2
Chromosome Location
Chr. 3: 146149796 - 146195514 on Build GRCm38
UniGene ID
Mm.116862
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005846.2 NP_001005846.1 3, 4 486
NM_026656.4 NP_080932.2 3, 4 514
GenBank mRNA AF503575.1 - 3, 4 503
AK014467.1 - 3, 4 417
AK019454.1 - 3, 4 383
AK150446.1 - 3, 4 482
AK150547.1 - 3, 4 482
AK152067.1 - 3, 4 482
AK152943.1 - 3, 4 482
AK153290.1 - 3, 4 482
AK159172.1 - 3, 4 514
AY083532.1 - 2, 3 332
BC029847.1 - 3, 4 363 3967456
CJ134616.1 - 3, 4 486

View Full Product Details