Gene Transcripts

Gene Symbol
Glod5
Entrez Gene ID
69824
Gene Name
glyoxalase domain containing 5
Gene Aliases
2010001H14Rik, AV061890
Chromosome Location
Chr. X: 8004200 - 8018492 on Build GRCm38
UniGene ID
Mm.279756
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027227.2 NP_081503.1 3 235
GenBank mRNA AK008008.1 - 3 238
BC061027.1 - 3 202 30312868

View Full Product Details