Gene Transcripts

Gene Symbol
Smap2
Entrez Gene ID
69780
Gene Name
small ArfGAP 2
Gene Aliases
1810031K02Rik, AA408614, Smap1l
Chromosome Location
Chr. 4: 120968317 - 121017289 on Build GRCm38
UniGene ID
Mm.271819
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133716.3 NP_598477.2 3 669
GenBank mRNA AK145042.1 - 3 654
BC031581.1 - 3 658
BC032217.1 - 3 645
BC052413.1 - 3 669 5720177

View Full Product Details