Gene Transcripts

Gene Symbol
Fbxo28
Entrez Gene ID
67948
Gene Name
F-box protein 28
Gene Aliases
4833428J17Rik, 5730505P19Rik, D1Ertd578e, Fbx28, mKIAA0483
Chromosome Location
Chr. 1: 182313102 - 182341606 on Build GRCm38
UniGene ID
Mm.30737
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175127.2 NP_780336.1 3 424
GenBank mRNA AK076211.1 - 3 430
AK077653.1 - 3 437
AK122295.1 - 3 409
AK162091.1 - 3 393
AK162188.1 - 3 305
BC055458.1 - 2 126
BC094542.1 - 3 424 6408744

View Full Product Details