Gene Transcripts

Gene Symbol
Pdrg1
Entrez Gene ID
68559
Gene Name
p53 and DNA damage regulated 1
Gene Aliases
1110004D19Rik, AU020945, AV006589, Pdrg
Chromosome Location
Chr. 2: 153008890 - 153015383 on Build GRCm38
UniGene ID
Mm.12746
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178939.2 NP_849270.1 3 281
GenBank mRNA AK003410.1 - 3 283
AK145939.1 - 3 326
AY286302.1 - 3 262
BC031699.1 - 3 281 4020218

View Full Product Details