Gene Transcripts

Gene Symbol
Trem1
Entrez Gene ID
58217
Gene Name
triggering receptor expressed on myeloid cells 1
Gene Aliases
Chromosome Location
Chr. 17: 48232367 - 48246924 on Build GRCm38
UniGene ID
Mm.248352
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021406.5 NP_067381.1 2 147
GenBank mRNA AF241219.1 - 2 102
AK089439.1 - 2 116
AK171753.1 - 2 116
BY203861.1 - 2 129

View Full Product Details