Gene Transcripts

Gene Symbol
Sp5
Entrez Gene ID
64406
Gene Name
trans-acting transcription factor 5
Gene Aliases
Chromosome Location
Chr. 2: 70474923 - 70477729 on Build GRCm38
UniGene ID
Mm.423397
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022435.2 NP_071880.1 2 1892
GenBank mRNA AF279479.1 - 2 1649
AK036234.1 - 2 1892

View Full Product Details