Gene Transcripts

Gene Symbol
Alg14
Entrez Gene ID
66789
Gene Name
asparagine-linked glycosylation 14
Gene Aliases
5430428G01Rik, AI854024
Chromosome Location
Chr. 3: 121291716 - 121362980 on Build GRCm38
UniGene ID
Mm.269881
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024178.2 NP_077140.1 3 345
GenBank mRNA AK011733.1 - 3 391
AK017354.1 - 2 46
BC002278.1 - 3 345 3496897

View Full Product Details